Learn More
Medchemexpress LLC OH-Glu-Val-Cit-PAB-MMAE | 1895916-23-0 | 99.0% | 1237.53 | C63H100N10O15 | 10 MG

Supplier: Medchemexpress LLC HY14824510MG
OH-Glu-Val-Cit-PAB-MMAE is a drug-linker conjugate combining a cleavable peptide linker (OH-Glu-Val-Cit-PAB) with the tubulin inhibitor monomethyl auristatin E (MMAE). It is used as a building block in the synthesis of antibody-drug conjugates (ADCs) for research applications.
- Cleavable peptide linker compatible with ADC synthesis.
- Contains monomethyl auristatin E (MMAE), a potent tubulin inhibitor.
- CAS number 1895916-23-0 and molecular weight 1237.53.
- High purity (99.0%) suitable for research use.
- Soluble in DMSO at 180 mg/mL; may require sonication.
- Storage: solid sealed at -20°C; in solvent store at -80°C up to 6 months.
- Available in small research quantities, such as 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.