Learn More
Research Products International Corp OHAUS Explorer Balance, EX423
SDP
Supplier:  Research Products International Corp 401001
Encompass
The OHAUS Explorer Series of analytical balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances and all of their intelligent features simplify even the most complex laboratory measurements.
Features such as four-level user management with password protection and a non-editable system log plus a high level of configuration make Explorer balances well suited for regulated applications.
A detachable terminal with a large color touchscreen, programmable IR sensors, automatic doors, and a frameless draftshield make Explorer analytical balances easy and convenient to use.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-197-6822