Learn More
Showa Denko America Inc OHpak SB-802 HQ
Supplier:  Showa Denko America Inc F6429100
Encompass_Preferred
Polyhydroxymethacrylate packed columns for aqueous SEC (GFC) analysis, supports a wide range of molecular weight sample analysis, the eluent can be replaced with DMF (except SB-802 HQ and SB-807 HQ), enabling the analysis of polar polymers, SB-804 and SB-805 used for accurate MW determination of gelatin by PAGI method, USP L38 and L39 listed, SB-802 and 802.5 HQ fulfill USP L25 requirements, SB-803 HQ fulfills USP L37 requirements
Catalog No. 50-161-3270