Learn More
Selleck Chemical LLC OICR-8268 E5839-5MG
SDP
Supplier:  Selleck Chemical LLC E58395MG
Encompass_Preferred
OICR-8268 is a potent reversible and in-cell active DCAF1 ligand It significantly binds to DCAF1 WDR domain with an Kd value of 38 nM It serves as a promising starting point for developing chemical handles for DCAF1-based PROTACs targeting selective protein degradation as well as cancer therapeutics
Catalog No. 50-000-41400