Learn More
Selleck Chemical LLC OICR-9429 5mg 1801787-56-3
SDP
Supplier:  Selleck Chemical LLC S78335MG
Encompass_Preferred
OICR-9429 is a potent antagonist of the interaction of WDR5 with peptide regions of MLL and Histone 3 and reduces viability of acute myeloid leukemia cells in vitro. It binds to WDR5 with high affinity (KD = 93 ¿ 28 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1339982