Learn More
ATCC Oidiodendron tenuissimum (Peck) Hughes, ATCC 22491 freeze-dried fungal research culture (BSL 1)
Supplier:  ATCC 22491
Encompass
Oidiodendron tenuissimum (Peck) Hughes is a freeze-dried fungal research culture supplied for laboratory research. The culture is intended for research use only and is not for therapeutic, diagnostic, or consumption purposes.
  • Freeze-dried format for long-term storage.
  • Type strain isolated from oak-pine forest soil.
  • Origin: New York, United States.
  • Biosafety level 1 organism.
  • Recommended growth medium: malt agar.
  • Incubation temperature: 24°C.
  • Environmental isolate suitable for mycological research.
  • Viability warranted for 30 days when stored and handled according to product instructions.
Catalog No. NC3722067