Learn More
Chemglass Life Sciences Chemglass Oil Bath, 350 mL, 100 x 50 mm (CG-1100-01)
SDP
Supplier:  Chemglass Life Sciences CG110001
Encompass_Preferred
Chemglass CG-1100-01 low-form oil bath is a 350 mL stainless steel heating dish designed for rapid, uniform heating of flasks, beakers, and other laboratory glassware. It accepts magnetic stirring and includes accessories for mounting and temperature sensing; an external temperature controller is required to maintain a set temperature.
  • 350 mL capacity.
  • 100 x 50 mm inner diameter x height.
  • Stainless steel construction.
  • 120 V, 400 W heating band.
  • Fits round bottom flasks 300 mL and smaller.
  • Includes lid, support block for rod attachment, and thermocouple clip.
  • Requires external temperature controller to maintain set temperature.
  • Compatible with magnetic stirring.
Catalog No. 50-273-3598