Learn More
Alkali Scientific Oil Centrifuge Tubes, 100 mL, 3.3 Borosilicate Glass, Blue Graduations, 8 Inch Tube (200x36mm), 10/Case
SDP
Supplier:  Alkali Scientific OT08CS
Encompass_Preferred
These oil centrifuge tubes are designed for the testing of petroleum products for water and sediment content in accordance with common laboratory and industry test protocols. Manufactured from premium 3.3 borosilicate glass, the tubes provide excellent chemical resistance, thermal stability, and durability for repeated laboratory use. Each tube is graduated to 100 mL with durable blue enamel graduations for clear and accurate volume measurement. A white printed marking area allows for easy sample identification and labeling. The tubes are reusable and autoclavable, supporting routine cleaning and sterilization without compromising performance. Available in 6 inch (short cone) and 8 inch configurations, these centrifuge tubes are compatible with most standard centrifuge equipment used in petroleum, fuel, lubricant, and quality control laboratories. Available Variants OT06 – 6 Inch Short Cone Oil Centrifuge Tube Capacity: 100 mL Dimensions: 165 mm length × 45 mm diameter Packaging: 1/Pa
Catalog No. 50-006-62798