Learn More
Diagnostic Biosystems OIL RED O Oil Red O
Supplier:  Diagnostic Biosystems KT025
Encompass_Preferred
Oil Red O Stain Kit (For Fat) is intended for use in the histological visualization of fat cells and neutral fat. This kit may be used ONLY on frozen tissue sections, fresh smears, or touch preps.
Catalog No. 50-240-7492