Learn More
Selleck Chemical LLC Oil Red O S4498-100mg
SDP
Supplier:  Selleck Chemical LLC S4498100MG
Encompass_Preferred
Oil red O (ORO) is a commonly used dye for the tissue staining which recognizes nitroglycerin and cholesterol esters Oil red O can be fluorescently lighted up upon meeting dsDNA with emission peak localized at 600 nm although it almost does not fluoresce by itself
Catalog No. 50-311-0473