Learn More
Chemglass Life Sciences Oil, Vacuum Pump, Duo-Seal, Quart
SDP
Supplier:  Chemglass Life Sciences AF036010
Encompass_Preferred
Oil, Vacuum Pump, Duo-Seal, Quart. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Duo-Seal oil is for use with belt driven (low RPM) AF-0350 pumps.
Catalog No. 50-872-675