Learn More
Medchemexpress LLC Olanzapine-d3 | 786686-79-1 | 99.1% | 315.45 g·mol⁻¹ | C17H17D3N4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14541S5MG
Encompass_Preferred
Olanzapine-d3 is the deuterium-labeled analogue of olanzapine used as an internal standard and analytical reference for quantitative bioanalysis. It enables mass-differentiated detection in LC-MS/MS workflows, improving accuracy and precision when measuring olanzapine in biological samples.
  • Deuterated internal standard for LC-MS/MS assays.
  • Enables mass-differentiated detection for accurate quantitation.
  • High chemical purity (99.1%) for reliable analytical performance.
  • Molecular weight 315.45 g·mol⁻¹ and formula C17H17D3N4S.
  • Suitable for use in plasma, serum, and other biological matrices.
  • Supplied as a small solid aliquot suitable for calibration and method development.
Catalog No. 50-002-21635