Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olanzapine N-oxide | 174794-02-6 | 98.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11939310MG

Encompass_Preferred

Olanzapine N-oxide is a drug metabolite and research compound used to study the metabolism and analytical profiling of olanzapine. It is supplied as a solid powder with supporting characterization data for laboratory and in vitro research.

  • Provided as a solid powder for analytical and in vitro studies.
  • High purity (98.0%) suitable for analytical applications.
  • Available in small-scale sizes, including 10 mg.
  • Characterized by COA, HNMR, LCMS, and RP-HPLC documentation.
  • Stable when stored under recommended conditions (powder: -20°C or 4°C).

Catalog No. 50-002-31783


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.