Learn More
Sigma Aldrich Fine Chemicals Biosciences ImprintR RNA Immunoprecipi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences RIP12RXN
Encompass
RNA immunoprecipitation (RIP) is a very powerful procedure for the study of RNA binding proteins (RBPs) and their RNA targets in ribonucleoprotein (RNP) complexes. Antibodies raised against specific RBPs are used to coprecipitate RNPs i.e. the RBP along with its RNA partner. The RNA can then be identified by next generation sequencing or if testing for a specific RNA by RT-PCR.
Catalog No. 50-290-7224