Learn More
Medchemexpress LLC Olanzapine | 132539-06-1 | 99.98% | C17H20N4S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14541R
Encompass_Preferred
Olanzapine is an atypical antipsychotic and an analytical standard intended for research and analytical applications. It is a selective, orally active monoaminergic antagonist with high affinity binding to serotonin, dopamine, muscarinic, and adrenergic α1 receptors. This compound serves as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Selective, orally active monoaminergic antagonist
  • Binds to serotonin H1, 5HT2A/2C, 5HT3, 5HT6 receptors
  • Binds to dopamine D1-4 receptors
  • Binds to muscarinic M1-5 receptors
  • Binds to adrenergic α1 receptors
  • Functions as an analytical reference standard
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-53243