Learn More
Medchemexpress LLC HY-10162 2g , Olaparib AZD2281;KU0059436 CAS:763113-22-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY101622G
Encompass_Preferred
Medchemexpress, HY-10162 2g Olaparib AZD2281;KU0059436 CAS:763113-22-0 Formula:C24H23FN4O3 IC50: 5/1 nM (PARP-1/2) Purity:98% Olaparib is a potent PARP inhibitor with IC 50 of 5 and 1 nM for PARP-1 and PARP-2 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2272