Learn More
Selleck Chemical LLC Olaparib (AZD2281) 10mM/1mL 763113-22-0 Ku-0059436
SDP
Supplier:  Selleck Chemical LLC S106010MM/1ML
Encompass_Preferred
Olaparib (AZD2281, KU0059436) is a selective inhibitor of PARP1/2 with IC50 of 5 nM/1 nM in cell-free assays, 300-times less effective against tankyrase-1. Olaparib induces significant autophagy that is associated with mitophagy in cells with BRCA mutations. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0968989