Learn More
Apexbio Technology LLC Olaparib (AZD2281, Ku-0059436), 500mg. Cas: 763113-22-0 MFCD: MFCD13185161. Potent PARP1/PARP2 inhibitor
SDP
Supplier:  Apexbio Technology LLC A4154500
Encompass_Preferred
Olaparib (4-(3-4-fluorophenyl) methyl-1(2H)-one), as known as AZD2281 or KU0059436, is a novel, selective and potent inhibitor of both poly adenosine diphosphate-ribose polymeras-1 (PARP-1) and poly adenosine diphosphate-ribose polymeras-2 (PARP-2). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4894