Learn More
Cayman Chemical OlaparIb-d4 1mg
SDP
Supplier:  Cayman Chemical 90027001 MG
Encompass_Preferred
An internal standard for the quantification of olaparib by GC- or LC-MS
Catalog No. 50-289-4657