Learn More
Medchemexpress LLC Olaparib-d8 | MFCD13185161 | 99.6% | 442.51 g·mol^-1 | C24H15D8FN4O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10162S110MG
Encompass_Preferred
Olaparib-d8 is the deuterium-labeled analog of olaparib, a potent PARP1/PARP2 inhibitor. This isotopically labeled compound is intended for research use, especially as an internal standard in LC-MS/MS quantitation and tracer studies. The compound has formula C24H15D8FN4O3 and a molecular weight of 442.51 g·mol⁻1.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • High isotopic purity suitable for quantitative analyses.
  • Validated by RP-HPLC and LC-MS for purity assessment.
  • Provided as a small-quantity research reagent (10 mg).
  • Suitable for tracer and pharmacokinetic studies.
Catalog No. 50-002-46999