Learn More
Medchemexpress LLC Olcegepant (BIBN-4096) | 204697-65-4 | 99.6% | 869.65 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY10095
Encompass_Preferred
Olcegepant (BIBN-4096) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor. It exhibits high affinity for the human CGRP receptor and can reverse CGRP-mediated vasodilation. Olcegepant has been studied for its role in inhibiting neurogenic vasodilation and reducing mechanical allodynia in animal models of migraine.
  • Potent and selective non-peptide CGRP1 receptor antagonist
  • IC50 of 0.03 nM and Ki of 14.4 pM for human CGRP
  • Exhibits higher affinity for human CGRP receptor than endogenous ligand CGRP
  • Reverses CGRP-mediated vasodilation in human cerebral vessels
  • Inhibits neurogenic vasodilation in animal models of migraine pathophysiology
  • Reduces mechanical allodynia in CCI-ION rats
Catalog No. 50-003-64433