Learn More
Apexbio Technology LLC OLDA 105955-11-1 50mg
SDP
Supplier:  Apexbio Technology LLC B681450
Encompass_Preferred
N-oleoyldopamine (OLDA CAS 105955-11-1) is a synthetic lipid amide formed by the conjugation of dopamine to oleic acid OLDA is recognized as an endogenous ligand for the transient receptor potential vanilloid 1 (TRPV1) channel where it acts as an agonist to induce channel activation Through TRPV1 modulation OLDA has been shown to influence physiological processes such as nociception and neurogenic inflammation This compound is frequently utilized in neuropharmacological research to investigate TRPV1-mediated signaling pathways and to explore endovanilloid function in pain sensation and related neurological disorders
Catalog No. 50-000-91229