Learn More
Medchemexpress LLC Olean-12-en-28-oic acid, 2,3,6,23-tetrahydroxy-, (2α,3β,4α,6β)- | 564-13-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN765210MG
Encompass_Preferred
Terminolic acid is a pentacyclic triterpene glycoside and an antibacterial agent, isolated from Combretum racemosum. It functions by inhibiting proinflammatory cytokines, specifically binding to the receptor active sites of IL-1β and IL-6, and reducing IL-8. This compound also exhibits antibacterial activity against various bacteria and is explored in colon cancer research.
  • Pentacyclic triterpene glycoside
  • Antibacterial agent
  • Isolated from Combretum racemosum
  • Inhibits proinflammatory cytokines IL-1β and IL-6
  • Reduces IL-8 production
  • Shows antibacterial activity against Staphylococcus aureus, Escherichia coli, and Enterococcus faecalis
  • Explored in colon cancer research
Catalog No. 50-004-18754