Learn More
Medchemexpress LLC Oleandrin | 465-16-7 | MFCD26960889 | 99.9% | 576.72 g·mol⁻1 | C32H48O9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY137195MG
Encompass_Preferred
Oleandrin is a cardiac glycoside used as a research reagent and analytical standard; it acts as a potent inhibitor of Na+/K+-ATPase and is applied in studies of ion transport, signal transduction, and cytotoxicity.
  • Potent Na+/K+-ATPase inhibitor with reported IC50 of 620 nM.
  • High reported purity (≈99.9%) suitable as an analytical standard.
  • Molecular formula C32H48O9 and molecular weight 576.72 g·mol⁻1.
  • Supplied in small research quantities for laboratory assays.
  • Useful in biochemical assays, pharmacology, and mechanism-of-action studies.
Catalog No. 50-002-21891