Learn More
Cayman Chemical OleIc ACDhydroxy Oleoyl Leu
SDP
Supplier:  Cayman Chemical 9004332100 UG
Encompass_Preferred
An internal standard for the quantification of oleic acid-hydroxy oleoyl leucine by GC- or LC-MS
Catalog No. 50-289-2215