Learn More
Cayman Chemical OleIc AcId-13C 10mg
SDP
Supplier:  Cayman Chemical 2786910 MG
Encompass_Preferred
An internal standard for the quantification of oleic acid by GC- or LC-MS
Catalog No. 50-293-7602