Learn More
Medchemexpress LLC Oleic acid-13C | 82005-44-5 | 100.0% | 283.45 | C1713CH34O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN1446S10MG
Encompass_Preferred
Oleic acid-13C is the 13C-stable isotope labeled form of oleic acid (9-cis-octadecenoic acid) supplied for research use as an analytical tracer or standard. It is used in biochemical and metabolic studies and is reported to modulate Na+/K+ ATPase activity.
  • 13C stable isotope labeled oleic acid.
  • High isotopic and chemical purity (99.99%).
  • Suitable as an analytical tracer or internal standard for metabolic studies.
  • Available in multiple small-mass packages, including 10 mg.
  • Molecular weight 283.45 g/mol; CAS number 82005-44-5.
Catalog No. 50-002-95708