Learn More
Cayman Chemical OleIc AcId-13C5 1mg
SDP
Supplier:  Cayman Chemical 90040891 MG
Encompass_Preferred
An internal standard for the quantification of oleic acid by GC- or LC-MS
Catalog No. 50-289-5668