Learn More
Cayman Chemical ANTIBODY PPZ-A10 5mg
SDP
Supplier:  Cayman Chemical 90041445 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of siRNA and mRNA in vitro and in vivo; intraperitoneal administration of LNPs containing PPZ-A10 and encapsulating an mRNA reporter preferentially accumulates in hepatic Kupffer cells and splenic macrophages in mice
Catalog No. 50-289-5922