Learn More
Medchemexpress LLC Oleic acid alkyne | 151333-45-8 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1271121MG
Encompass_Preferred
Oleic acid alkyne is oleic acid with an acetylene group. The terminal alkyne group facilitates click chemical ligation reactions, enabling the introduction of fluorescent or biotin-labeled oleic acid. It can also be hydroxylated by a microsomal cytochrome P-450-dependent system (ω-OAH), allowing for analysis of its metabolism and biological activity.
  • Features a terminal alkyne group for click chemistry
  • Supports fluorescent or biotin labeling of oleic acid
  • Applicable for analyzing metabolism and biological activity
  • Can be hydroxylated by a microsomal cytochrome P-450-dependent system (ω-OAH)
Catalog No. 50-003-42657