Learn More
Medchemexpress LLC Oleic acid-¹³C | 82005-44-5 | 283.45 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN1446S1MG
Encompass_Preferred
Oleic acid-13C is a 13C labeled Oleic acid, an abundant monounsaturated fatty acid that acts as a Na+/K+ ATPase activator. This compound serves as a valuable tool for quantitative analysis in various scientific applications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-14664