Learn More
Medchemexpress LLC Oleic acid-d2 | 5711-29-5 | 284.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN1446S150MG
Encompass_Preferred
Oleic acid-d2 is the deuterium-labeled version of Oleic acid, an abundant monounsaturated fatty acid and a Na+/K+ ATPase activator. This product is intended for research use only.
  • Deuterium labeled Oleic acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15642