Learn More
Cayman Chemical Oleic Acid ethyl ester
SDP
Supplier:  Cayman Chemical 100082011 G
Encompass_Preferred
Oleic acid ethyl ester is a neutral, more lipid-soluble form of oleic acid.
Catalog No. 50-288-8714