Learn More
U.S. Pharmacopeia Oleic Acid, 112-80-1, MFCD00064242, 1g
Supplier:  U.S. Pharmacopeia 1478130
Encompass
Molecular formula C18H34O2, Molecular weight 282.46, Melting Point 37.4 - 60.8 °F (3 - 16 °C), Boiling Point 680 °F (360 °C), Flash point 372.2 °F (189.0 °C) Closed Cup, Synonyms: 9-Octadecenoic Acid
Catalog No. NC9588352