Learn More
Medchemexpress LLC Oleic acid sodium | 143-19-1 | 304.44 | 1 ML
Supplier: Medchemexpress LLC HYN1446B10MM/1ML
Sodium oleate is an abundant monounsaturated fatty acid sodium and functions as a Na+/K+ ATPase activator.
- Functions as a Na+/K+ ATPase activator.
- Common monounsaturated fatty acid in human adipocytes and tissues.
- Can influence cell proliferation and migration in cancer cells.
- High purity of 99.99%.
- Appears as a white to off-white solid.