Learn More
Cayman Chemical Oleic Acidbiotin
SDP
Supplier:  Cayman Chemical 100106241 MG
Encompass_Preferred
An affinity probe designed to allow oleic acid to be detected in complexes with protein binding partners such as fatty acid binding proteins
Catalog No. 50-288-8436