Learn More
Medchemexpress LLC Oleic-DBCO | 2279951-78-7 | 98.0% | C36H48N2O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14129310MG
Encompass_Preferred
Oleic-DBCO is a DBCO-functionalized oleic acid derivative used as an alkyl chain-based PROTAC linker and a click chemistry reagent. The DBCO (dibenzocyclooctyne) moiety allows strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules, enabling bioorthogonal conjugation for chemical biology and PROTAC synthesis. The compound is provided as a high-purity powder with specified storage conditions for stability.
  • Alkyl chain linker suitable for PROTAC synthesis.
  • Enables copper-free click chemistry via strain-promoted alkyne-azide cycloaddition.
  • High purity: 98.0%.
  • Molecular formula C36H48N2O2 and molecular weight 540.78.
  • Stable as a powder at -20°C for up to 3 years.
  • Available in small milligram-scale quantities for research use.
Catalog No. 50-002-59452