Learn More
Medchemexpress LLC Oleoyl serotonin | 1002100-44-8 | MFCD18428060 | 99.0% | 440.66 | C28H44N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1098415MG
Encompass_Preferred
Oleoyl serotonin is a small-molecule TRPV1 antagonist used in laboratory research. The compound shows a reported IC50 of 2.57 μM against human TRPV1 and is provided at high purity for experimental pharmacology studies. It is intended for research use only.
  • TRPV1 antagonist with reported IC50 of 2.57 μM for human TRPV1.
  • High purity, 99.0%.
  • Molecular weight 440.66; formula C28H44N2O2.
  • Suitable for in vitro and in vivo research applications.
  • Provided in small research-scale quantities (for example, 5 mg).
Catalog No. 50-002-64691