Learn More
Cayman Chemical Oleoyl SerotonIn 50mg
SDP
Supplier:  Cayman Chemical 900062950 MG
Encompass_Preferred
A hybrid molecule patterned after N-arachidonoyl serotonin; inhibits capsaicin-induced TRPV1 channel activation (IC50 = 2.57 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM)
Catalog No. 50-289-4530