Learn More
Cayman Chemical Oleoyl SerotonIn-d17 5mg
SDP
Supplier:  Cayman Chemical 90006945 MG
Encompass_Preferred
An internal standard for the quantification of oleoyl serotonin by GC- or LC-MS
Catalog No. 50-289-3920