Learn More
Cayman Chemical OleoylCoenzyme AsodIM sal 10mg
SDP
Supplier:  Cayman Chemical 3538410 MG
Encompass_Preferred
An acyl-CoA thioester; a substrate for ACAT-1, ACAT-2, ACAD-9, MBOAT1, and MBOAT2; binds the FadR promoter in E. coli (Ki = 0.005 µM); activates SUR1/Kir6.2 in Xenopus oocytes at 1 µM
Catalog No. 50-291-3389