Learn More
Apexbio Technology LLC Oleuropein 32619-42-4 20mg
SDP
Supplier:  Apexbio Technology LLC N186320
Encompass_Preferred
Oleuropein (CAS 32619-42-4) is a phenolic glycoside compound found in olive leaves and oil It is designed to modulate cellular redox balance and apoptotic pathways thereby influencing oxidative stress inflammation and cell survival Oleuropein exerts its biological activity primarily through antioxidant action inhibition of transcriptional activity and induction of apoptosis via the p53-dependent pathway as well as regulation of related genes In cell-based studies oleuropein demonstrates inhibition of aromatase and induces apoptosis in breast cancer cells Based on these pharmacological properties oleuropein holds research potential in cancer biology inflammation atherosclerosis and studies of cellular antioxidant responses
Catalog No. 50-000-91655