Learn More
Medchemexpress LLC HY-N0292 5mg Medchemexpress, Oleuropein CAS:32619-42-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02925MG
Encompass_Preferred
Medchemexpress, HY-N0292 5mg Oleuropein CAS:32619-42-4 Oleuropein, found in olive leaves and oil, exerts antioxidant, anti-inflammatory and anti-atherogenic effects through direct inhibition of PPARγ transcriptional activity. Oleuropein induces apoptosis in breast cancer cells via the p53-dependent pathway and through the regulation of Bax and Bcl2 genes. Oleuropein also inhibits aromatase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0250