Learn More
Medchemexpress LLC Oleuropeinic acid | 96382-90-0 | 1 MG

Supplier: Medchemexpress LLC HYN68751MG
Oleuropeinic acid is present initially in olive tissues or formed by thermal treatment, possibly by the oxidation of oleuropein. It is an antioxidant-soluble fiber.
- Purity: 97.80%
- Molecular weight: 570.50
- Formula: C25H30O15
- Appearance: Solid
- Color: White to off-white
- Soluble in DMSO (≥ 100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.