Learn More
Medchemexpress LLC Oleuropeinic acid | 96382-90-0 | 10 MG

Supplier: Medchemexpress LLC HYN687510MG
Oleuropeinic acid is an antioxidant-soluble fiber that is present in olive tissues or formed by the thermal treatment of oleuropein.
- Antioxidant-soluble fiber
- Derived from olive tissues
- Classification includes terpenoids, iridoids, phenols, and polyphenols
- Store at 4°C in sealed conditions, away from moisture and light
- Solubility in DMSO: ≥ 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.