Learn More
Medchemexpress LLC Oleuropeinic acid | 96382-90-0 | 5 MG

Supplier: Medchemexpress LLC HYN68755MG
Oleuropeinic acid is found in olive tissues and can also be formed through the thermal oxidation of oleuropein. It is recognized as an antioxidant-soluble fiber.
- Acts as an antioxidant-soluble fiber
- Purity of 97.80%
- Molecular weight of 570.50
- Appears as a white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.