Learn More
Medchemexpress LLC Oleyl alcohol | 143-28-2 | 98.0% | C18H36O | 100 MG

Supplier: Medchemexpress LLC HY141573100MG
Oleyl alcohol is an active compound for various studies. It is a colorless to light yellow liquid with a density of 0.8489 g/cm³ and a molecular weight of 268.48. This product is for research use only and has a purity of 98.0%.
- Active compound
- Colorless to light yellow liquid appearance
- Density: 0.8489 g/cm³
- Molecular weight: 268.48
- Purity: 98.0%
- Chemical formula: C18H36O
- Shipping: Room temperature (continental US)
- Storage (pure form): -20°C for 3 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Solubility in DMSO: ≥ 30 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.