Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleyl alcohol | 143-28-2 | 98.0% | C18H36O | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY141573

Encompass_Preferred

Oleyl alcohol is an active compound used for various studies. It is a colorless to light yellow liquid with a density of 0.8489 g/cm³ and a molecular weight of 268.48.

  • Active compound for various studies
  • Colorless to light yellow liquid
  • Density of 0.8489 g/cm³
  • Molecular weight of 268.48

Catalog No. 50-003-52822


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.