Learn More
Medchemexpress LLC Oleyl alcohol | 143-28-2 | 98.0% | C18H36O | 500 MG

Supplier: Medchemexpress LLC HY141573500MG
Oleyl alcohol is an active compound intended for research use only in laboratory settings. This chemical reagent is stable under recommended storage conditions and has not been fully validated for medical applications.
- Active compound for various studies
- Stable under recommended storage conditions
- For research use only
- Not intended for human use
- Tested and complies with specifications
- Appearance: Colorless to light yellow liquid
- Purity: 98.0% (GC)
- Molecular formula: C18H36O
- Molecular weight: 268.48
- CAS number: 143-28-2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.