Learn More
Cayman Chemical Oleyl Anilide Inhibitors
SDP
Supplier:  Cayman Chemical 100065295 MG
Encompass_Preferred
An ACAT inhibitor (IC50 = 26 µM); induces splenomegaly in C57BL/6 mice and leads to weakness, weight loss, cachexia, and death in A/J mice with increases in serum IgG and IgM levels in both strains; has been found in aniline-denatured rapeseed cooking oil, which has been associated with toxic oil syndrome
Catalog No. 50-288-9913